Axygen® Deep Well Plates, 384 Well, 120 and 240 µl, Corning
Supplier: Corning
P-384-240SQ-C-SI
47745-406CS 987.12 USD
47745-406
Axygen® Deep Well Plates, 384 Well, 120 and 240 µl, Corning
Microplates Deep Well Microplates
PP. Used for sample collection, cell culture and long-term storage.
  • Square wells
  • Excellent chemical resistance
  • Temperature resistant down to −80 °C
  • Can withstand centrifugation up to 4000 ×g
Two types of sealing mats are suitable for deep 384-well microplates:
AxyMat™ sealing mats are designed for high throughput screening and storage applications. Made of silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be re-used. AxyMat™ sealing mats are pierceable and self-sealing, ideal for automated processes, and can be used in sub-zero environments.
ImpermaMat™ sealing mats are chemically resistant. They are made for use in assays involving strong solvents such as DMSO and similarly harsh solvents. Re-usable. They feature a tight seal to the plate so there is no sample evaporation or well-to-well evaporation. Usable within a wide temperature range: from −80 up to +121 °C.