Portable Digital Megohmmeter, ACL
Supplier: ACL Staticide
ACL 800
47744-792EA 1958.99 USD
47744-792
Portable Digital Megohmmeter, ACL
Electrical Meters Digital Ohmmeters
Ideal for measuring the resistance of packaging, smocks, anti-static mats, work surfaces, and flooring
Measures temperature, humidity, and electrical resistance/resistivity. Uses both internal and external probes. Measures resistance point-to-point from 1e3 to 1e13ohm. 10V and 100V test voltages.
Certifications: ESD Association Standards S4.1 and S7.1. European Standards IEC 61340-4-1, 61940-2-3, and 61340-4-5.