Corning® T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Corning
Supplier: Corning
354115
47743-642EA 759.12 USD
47743-642
Corning® T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Corning
Cell Culture Supplements and Additives
T-STIM with ConA (IL-2 culture supplement), rat, is used to promote proliferation and activation of T-cells and support high-titer HIV production by leukocytes.
Media used to promote proliferation and activation (PHA or ConA Supplemented Media) of T-cells and support high-titer HIV production by leukocytes.
Tested for ability to promote proliferation of an IL-2 dependent mouse cell line (CTLL) in serum-containing medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.