Nitri-Master® Gloves, Showa
Supplier: Showa
NM11-07 NM11-08 NM11-09 NM11-10 NM11-11
47742-406CS 760.13 USD
47742-406 47742-408 47742-410 47742-412 47742-414
Nitri-Master® Gloves, Showa
Gloves
Ergonomically designed forms to reduce fatigue and enhance comfort.
  • Gloves are unlined and feature a tractor tread grip
  • Flexible, low-modulus glove
  • Embossed grip
Nitri-Master® gloves are for manufacturing, chemical processing, petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications, and machine applications using cutting oils.
Certifications: 21 CFR / FDA compliant for direct food contact.