SHOWA 6781R Insulated Neoprene, Chemical Resistant Glove, 18", Showa
Supplier: Showa
6781R-06-10
47742-234CS 985.88 USD
47742-234
SHOWA 6781R Insulated Neoprene, Chemical Resistant Glove, 18", Showa
Gloves
Ideal for applications requiring protections from acids and chemicals.
  • 18" length
  • Neoprene coating resists acids and chemicals
  • Insulated for thermal protection
A glove used for protection from a wide range of chemicals. They provide elbow length protection from most acids.
Gloves also provide protection in hot and cold environments.