VWR® Spinbar® Fluted Octagonal Rare Earth Magnetic Stirring Bar
Supplier: VWR International
37174-6000
47735-602CS 1277.47 USD
47735-602
VWR® Spinbar® Fluted Octagonal Rare Earth Magnetic Stirring Bar
Stirrers Magnetic Stirring Bars
This bar’s high-energy magnetic material, combined with a fluted octagonal shape, produces exceptional power, although its length is only 50 mm (1.969") with a 21 mm (0.83") diameter.
  • Ideal for viscous solutions and efficient, high-speed stirring
  • Samarium Cobalt magnet encapsulated in FDA grade, USP Class VI Teflon® PTFE provides strong coupling with drive magnets
  • Recessed flutes and distinct edges of this eight-sided bar provide excellent surface area; generates strong turbulence so liquids are thoroughly mixed
  • Stability is not impaired in vessels with curved bottoms
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.