Refractive Index Liquids, Series AAA
Supplier: CARGILLE LABORATORIES INC.
1803X
47735-584EA 133.6 USD
47735-584
Refractive Index Liquids, Series AAA
Standards Refractive Index Standards
Refractive index liquids have a broad range of applications in diversified fields.
  • Slightly volatile
  • Colorless flurocarbon formulation
Adjustment: ±0.0002
Choose from Range nD 1.300-1.395
Ordering information: Required refractive index has to be specified with order.