Cannon Bath Cooler, 115 V, Cannon Instrument Company
Supplier: Cannon
9726-D05
47735-078EA 8691.24 USD
47735-078
Cannon Bath Cooler, 115 V, Cannon Instrument Company
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The compact and efficient Cannon Bath Cooler (CBC) uses an advanced technological design to maintain a liquid bath below room temperature
With the CBC, bath temperatures can be lowered up to 15°C below ambient. The CBC consists of a thermoelectric Cooling Unit and a separate power supply.