CT-2000 Series Constant Temperature Bath, 240 V, Cannon Instrument Company
Supplier: Cannon
9726-A35
47735-076EA 20033.82 USD
47735-076
CT-2000 Series Constant Temperature Bath, 240 V, Cannon Instrument Company
Baths Viscometer Baths
Convenient keypad operation and ASTM reliability at temperatures to 150°C make the CT-2000 the bath of choice for many users
  • 240 V model, not for use in USA
Microprocessor-controlled temperature regulation, on-screen temperature monitoring, and serial networking options make this digital bath an attractive and fully functional addition to the modern laboratory.
Caution: 240 V model, not for use in USA. Corresponding model with voltage suitable for use in US (47735-074) is also available, please see separate product page.