CT-2000 Series Constant Temperature Bath, 120 V, Cannon Instrument Company
Supplier: Cannon
9726-A30
47735-074EA 20033.82 USD
47735-074
CT-2000 Series Constant Temperature Bath, 120 V, Cannon Instrument Company
Baths Viscometer Baths
Convenient keypad operation and ASTM reliability at temperatures to 150°C make the CT-2000 the bath of choice for many users
Microprocessor-controlled temperature regulation, on-screen temperature monitoring, and serial networking options make this digital bath an attractive and fully functional addition to the modern laboratory.