Optic Prep™ Presaturated Wipes, ITW Chemtronics®
Supplier: ITW Chemtronics
CP410
47733-174CS 199.65 USD
47733-174
Optic Prep™ Presaturated Wipes, ITW Chemtronics®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
High quality premoistened lens-grade tissue for cleaning optical surfaces.
  • Safely clean and polish optical surfaces, lenses, photographic equipment, CRT screens, and instruments
  • Ideal for field service kits
  • Nonabrasive
Quickly remove dust, oils, and contaminants. Solvent evaporates quickly and leaves no residue. Compatible with most plastics, glass, and metals. Each individual wipe is hermetically sealed to prevent evaporation and maintain solvent effectiveness.