Flux-Off® Water-Soluble Flux Remover, ITW Chemtronics®
Supplier: ITW Chemtronics
ES1530
47732-656CS 35.71 USD
47732-656
Flux-Off® Water-Soluble Flux Remover, ITW Chemtronics®
Soldering Supplies Flux Removers
Proprietary blend of powerful cleaning solvents that is safe for use on water-sensitive components
Engineered to remove Type R, RMA, RA, and synthetic flux residues, as well as ionic and non-ionic soils. Ozone- and platic-safe. Dries quickly and leaves no residue. CFC/HCFC-free. Flammable. Aerosol spray