Konform® SR Conformal Coating, ITW Chemtronics®
Supplier: ITW Chemtronics
CTSR-1
47732-654EA 514.18 USD
47732-654
Konform® SR Conformal Coating, ITW Chemtronics®
Sprays
Formulated to provide maximum flexibility for extreme temperatures, this transparent coating protects both rigid and flexible printed circuit boards for the highest long-term protection in adverse operating conditions
UL listed.
Meets MIL-I-46058C, Type SR. Usable temperature range: –65° to 200°C (–85° to 390°F).