Konform® AR Conformal Coating, ITW Chemtronics®
Supplier: ITW Chemtronics
CTAR-12
47732-652CS 865.06 USD
47732-652
Konform® AR Conformal Coating, ITW Chemtronics®
Sprays
Konform® AR is ideal for insulating against high-voltage arcing and corona shorts
UL listed.
This acrylic conformal coating provides a hard, durable protective barrier against humidity, salt, corrosive vapors, and fungus for printed circuit boards and electronic assemblies. Protects and increases life of electronic assemblies. Meets MIL-I-46058C, Type AR. Usable temperature range: –59° to 132°C (–75° to 270°F).