Compact Stand for Overhead Stirrers, Caframo
Supplier: Caframo
A210
47731-189EA 327.02 USD
47731-189
Compact Stand for Overhead Stirrers, Caframo
Support Stands
The compact stand is ideal for a small work space or under a fume hood.
The stand base is composed of cast zinc-aluminum alloy coated with chemical resistant epoxy.
Ordering information: Includes the stand base and 24" x 5/8" dia., 304 stainless steel solid rod.