Electrochemical XS sensors
Supplier: Draeger
6809145
47729-628EA 911.93 USD
47729-628 13450-212
Electrochemical XS sensors
Data Loggers
Electrochemical XS sensors can be used for detecting many different gases and vapours. These sensors are equipped with an integrated EEPROM (Electrically Erasable Programmable Read-Only Memory).
  • Long service life and low operating costs
  • Excellent reliability and high measurement quality
  • Stable, longer calibration intervals
  • All relevant sensor data are stored in memory module
Ideal for monitoring of toxic gases and oxygen deficiency including hydrides, amines, organic vapours.
The sensitivity data of more than 12 different gases and vapours are stored in the XS OV sensor, so that multiple gas hazards can be measured by a single sensor. The plug & play function allows sensors to be swapped between devices, without, for example, having to recalibrate them.