United Scientific Two-Way Table Clamp
Supplier: United Scientific Supplies
2WYCLMP
470359-286EA 30.8 USD
470359-286
United Scientific Two-Way Table Clamp
Clamps General Purpose Clamps
Securely hold rods in two dimensions.
  • Hold rods vertically or horizontally
  • Cast aluminum, nickel-plated clamp
  • Easy to adjust, nickel-plated steel thumbscrews
  • Fits surfaces up to 3” thick
Two-way clamp and thumbscrews hold rods firmly in horizontal or vertical positions. For use with rods up to ¾" in diameter. The cast aluminum, nickel-plated clamp mounts onto tabletops or other surfaces up to 3” thick. Screw mechanisms are made of nickel-plated steel.
Ordering information:
Rods are not included.