Cartridge Casing Collector
Supplier: THE LYNN PEAVEY COMPANY
95582
470358-988PK 29.95 USD
470358-988
Cartridge Casing Collector
Educational Materials Forensics Educational Materials Forensics Supplies
No contact pickup.
  • Mixed package size to cover multiple calibers
  • Easily pick up casings without touching evidence
  • Developed by a crime scene investigator
The 'CCC' Cartridge Casing Collector is a nifty and unique mechanism to pick up spent cartridge casings found at crime scenes, without transferring any DNA. These almost indestructible CCC’s have a small enough tip to fit inside 0.22 caliber up to 0.45 caliber rounds. Simply pinch the no-slip grip tips together, insert into the opening, then slightly release the pressure. Collects casings instantly. For DNA purposes, they are individually packaged in hermetically sealed bags. A larger, more reusable Cartridge Casing Collector is available that can handle all casings up to and including 12 gauge shotgun shell sized evidence. Durable and reusable, but for DNA we would recommend pitching or packaging with the collected evidence. Combo pack includes 5 small and 2 large.