Four-Way Digital Soil Analyzer
Supplier: Luster Leaf
1885
470358-646CS 105.3 USD
470358-646
Four-Way Digital Soil Analyzer
Educational Materials Environmental Educational Materials Environmental Learning Activities
Determines proper moisture, pH, light, and temperature in soil samples.
  • Large and easy to read digital display
  • Probe length 5"
  • Four function switch
Ensure healthy vegetative growth with accurate, calibrated readings. This device measures light levels adjusted for optimal theoretical plant growth across all light sources, including variations in natural daylight. Calibrated to the USDA mean solar radiation table, it provides reliable, research-based light data.
Also features soil temperature measurement in both fahrenheit and celsius, with an easy-to-reference guide showing ideal germination and growth temperature ranges for a wide variety of plants.