Total Bacteria Slides
Supplier: Culture Media & Supplies
CM858MC
470358-622PK 70.65 USD
470358-622
Total Bacteria Slides
Slides Prepared Slides
Used to monitor microbial contamination.
  • Tests for the pressence of bacteria
  • Also known as a dip slide
  • No special equipment or training necessary
  • MS/HS grade level activity
Supplied with test instructions and results chart. Results in 24 to 48 hours after incubation. Resulting growth patterns are compared to the included chart to ascertain the level of microbial contamination.