Kidder Wooden House Frame STEM Build Kit
Supplier: KIDDER MFG. CO. LTD
80-50-W044
470358-038KT 16.4 USD
470358-038
Kidder Wooden House Frame STEM Build Kit
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Assemble a wooden house frame to explore architectural design and structural principles.
  • Encourages hands-on STEM learning
  • Demonstrates principles of triangle stability and rectangular balance
  • No cutting required; glue included
  • Includes online teacher resource guide
The Wooden House Frame STEM Build Kit offers students an engaging hands-on experience to explore architectural design and structural engineering principles. By assembling a model of a wooden house frame, learners gain insight into how triangle stability and rectangular balance contribute to the integrity of structures. This activity not only reinforces theoretical knowledge but also enhances problem-solving skills, making it an excellent addition to STEM education programs.
Application: Engineering, Mechanics