AquaSprouts Universal Pond Garden
Supplier: AQUASPROUTS, INC
9207
470357-832EA 199.95 USD
470357-832
AquaSprouts Universal Pond Garden
Educational Materials Biology Educational Materials Ecology Learning Activities
Elevate your gardening game by transforming any container into a thriving aquaponic or hydroponic environment.
  • Mountable grow bed is crafted from durable, weather-resistant materials, and designed to be easily mounted onto a variety of containers
  • Grow bed measures 29.5L×13W×8"H and has a capacity of approximately 10 gallons
  • Includes a submersible 160-gallon-per-hour pump for optimal water circulation and cleanliness (pump is suitable for both indoor and outdoor use)
  • Plant grow-bed can effectively treat up to 50 gallons of water, ensuring optimal filtration and nutrient balance for aquaponic or hydroponic systems
  • Control the flow rate of the grow bed and water level in the planter with customizable plumbing fittings
  • Expanded clay grow media replaces soil from traditional gardening and serves as the support for your plants; this media does not degrade
This compact and versatile grow-bed effortlessly converts any container with a 19" diameter or larger (such as flowerpots, barrels, stock tanks, and planters) to a flourishing ecosystem. Allowing you to repurpose containers you already own, reducing costs, and encouraging sustainable practices.
Whether you are a seasoned gardener or just starting out, you can now explore aquaponics or hydroponics with ease. It can also be used to create a tranquil water garden, or a simple fountain feature in a self-sustaining ecosystem.
Designed for simplicity and adaptability, this versatile grow-bed is easy to set up and fully customizable to fit your unique needs. It can be easily mounted onto the edge of a stock tank or other containers with the included bracket and hardware kit. The possibilities are endless with this all-in-one solution.