NPS® Polyurethane Task Chair with Arms, Black
Supplier: National Public Seating
6722HB-A
470357-628EA 510.15 USD
470357-628
NPS® Polyurethane Task Chair with Arms, Black
Chairs
Heavy duty 1¹/₄" thick polyurethane seat has a waterfall edge. Built on five 2" industrial grade caster wheels for portability.
  • Heavy duty 1¹/₄" thick polyurethane seat with waterfall edge
  • Polyurethane material is resistant to chemicals and punctures, and cleans easily
  • Features pneumatic height adjustment from 22 - 32"
  • Swivels 360 degrees for unlimited functionality
  • Integrated under seat spring system allows for control of backrest tilt degree with a turn of a knob
  • Built on five 2" industrial grade caster wheels for portability or install included mushroom glides for use in a stable environment
  • Convenient backrest handhold for maneuverability
  • Weight capacity 300 lbs.
  • featured with arms
The 6700 series kangaroo stool provides unique strength and function for your industrial seating demands, yet designed with beauty and grace in mind.  Heavy duty 1¹/₄" thick Polyurethane seat has a waterfall edge for plush comfort and functionality, and ribbed ventilation design allows air circulation while seated. Polyurethane material is resistant to chemicals and punctures, and cleans easily.  Includes accent trim for aesthetics. Featured with arms.
Certifications:
ANSI-BIFMA certified; MAS certified green.
Ordering information:
Ten-year warranty. Two-day lead time.