Fundamentals of Physics Force Table
Supplier: Wards
FN-FTBL VWRFOPBASE
470061-748EA 115.56 USD
470061-748 470357-594
Fundamentals of Physics Force Table
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
The Fundamentals of physics force table is economical and convenient.
  • Table marked with degrees
  • Three pulleys and hangers to simply demonstrate vector addition and equilibrant forces
  • Economical and convenient
  • Table diameter is 30 cm
It delivers the accuracy of a full unit but with a detachable base, it can be stored in half the space.
The table's support rod is designed to attach to the FUN stand base, which makes the Fundamentals of physics product line modular and easy to set up and use. It requires the FUN stand, available separately.
The table is marked with degrees and features three hanging, stainless steel masses.
Accessories information:
FUN stand base is required, but available separately.
Ordering information: FUN stand base is required but not included.