Hanna 1382 mg/L (ppm) TDS Calibration Solution, Hanna
Supplier: HANNA INSTRUMENTS
HI70032P
470357-560EA 39.3 USD
470357-560
Hanna 1382 mg/L (ppm) TDS Calibration Solution, Hanna
Standards Conductivity Standards
NIST traceable and contained in portable single-use sachets.
  • Temperature chart of the actual µS/cm value at various temperatures printed on each bottle
  • Convenient packaging for one time use
Each sachet has the lot number and expiration date stamped on it and is made of light block foil ensuring freshness each time one is opened.
Expiration date of 5 years from the date of manufacture for an unopened sachet.