Hanna 1413 µS/cm Conductivity Standard
Supplier: HANNA INSTRUMENTS
HI70031P
470357-558EA
39.3
USD
470357-558
Hanna 1413 µS/cm Conductivity Standard
Standards
Conductivity Standards
NIST traceable and contained in portable single-use sachets.
Temperature chart of the actual µS/cm value at various temperatures printed on each bottle
- Convenient packaging for one time use
Each sachet has the lot number and expiration date stamped on it and is made of light block foil ensuring freshness each time one is opened.
Ordering information:
Expiration date of five years from the date of manufacture for an unopened sachet.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)