Hanna General-Purpose Sachet Cleaning Solution
Supplier: HANNA INSTRUMENTS
HI700601P
470357-554EA
29.2
USD
470357-554
Hanna General-Purpose Sachet Cleaning Solution
Electrolytes
Electrode Storage Solutions
Formulated for pH electrodes with a variety of applications.
25 tests
- Designed for 20 ml sachets
- Specially formulated for cleaning electrodes
- Convenient, one-time use sachets ensure freshness of solution
Electrodes can become dirty from use and will produce inaccurate results even as they read correctly in a pH buffer. This cleaning solution eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement and stored incorrectly. Hanna suggests cleaning the bulb and junction of the electrode on a regular basis to ensure that the probe is always clean and to prevent any clogging of the junction.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)