ProBath® Water Bath
Supplier: Revolutionary Science
RS-PB-150
470357-538EA 705.9 USD
470357-538
ProBath® Water Bath
Baths Water Baths, Unstirred
9½" vertical clearance easily accommodates tall items such as 1 L flasks or beakers without removing the cover.
  • Ideal for DNA extraction, CRISPR, and heat shock
  • 300 series stainless steel chamber construction
  • Simple 2-button operation
  • Over and under temperature alarms
  • Two year warranty
The ergonomic design has a small footprint to fit in tight bench spaces.
Delivery information:
Supplied with an impact-resistant polycarbonate cover.