AquaSprouts® Seedling Biodome
Supplier: AQUASPROUTS, INC
6292
470357-454EA 119.65 USD
470357-454
AquaSprouts® Seedling Biodome
Testing Chambers Plant Growth Chambers
Get a jump on your gardening goals with the AquaSprouts® seed sprouting greenhouse.
  • Biodome comes with 32 AquaSprouts grow plugs
  • Removable seedling tray holds up to 72 AquaSprouts grow plugs
  • 18" LED grow light fits perfectly on humidity cover
  • Smart plug requires a 2.4 GHz wifi network
With this biodome, you can grow year-round microgreens, herbs, and starter plants indoors, giving delicate plants a head-start on root growth. By starting your plants indoors you can ensure that they get better purchase and better nutrient absorption once you move them into your garden bed. So why wait? Get started on your gardening goals today - no matter the weather outside.
The smart seed sprouting biodome features a manual on/off switch, as well as the ability to connect to your smart phone for customizable timing schedules. The built-in light channel and two adjustable vents allows you to optimize the growing environment for any plant.
Ordering information: Supplied with humidity cover.
Caution: Note: requires a 2.4 GHz wifi network.