Ward's® Euhderal Fluorite Pack
Supplier: Avantor
470357-344PK 29.99 USD
470357-344
Ward's® Euhderal Fluorite Pack
Specimens Mineral Specimens Crystals
Essential for teaching mineralogy and atomic structure.
  • Cubic crystal structure
  • Translucent
  • Ideal for small group learning
These crystals showcase the beauty and complexity of crystal formation, helping students understand the concepts of symmetry, geometry, and mineral properties. Their well-defined crystal shapes provide a hands-on way to explore topics such as mineralogy, atomic structure, and the conditions required for crystal growth. By studying fluorite, students can also engage in discussions about Earth's geology, the rock cycle, and the practical uses of minerals in everyday life.