Med Dimensions® Macaw Intubation Trainer
Supplier: MED DIMENSIONS, LLC
MD-E-A1031S
470357-270EA 446.68 USD
470357-270
Med Dimensions® Macaw Intubation Trainer
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Ready to take your avian veterinary skills to the next level?
  • Derived from Ct scans, with relative anatomy including soft tongue, flexible jaw and airway to syrinx
  • Ideal for practicing intubation
  • Ideal for use with 3.5 Et tube
  • Bisected body to visualize the airway
Aspiring avian or exotics vets, this simulator is your key to acing macaw intubation. The Macaw Intubation Standard replicates the unique anatomy of a cockatoo’s beak and airway. Improves hand-eye coordination and masters instrument control for smooth, efficient intubation. Reduces training time by learning faster and retaining skills more effectively. Impress instructors and colleagues by showcasing your skills and dedication to avian care.
Caution: This product is a non-medical device intended solely for educational purposes. It is not designed, manufactured, or intended for use in the diagnosis, treatment, cure, mitigation, or prevention of disease or for any other medical purpose.