Med Dimensions® Canine Thoracentesis Trainer
Supplier: MED DIMENSIONS, LLC
MD-E-C2023
470357-234EA 3445.6 USD
470357-234
Med Dimensions® Canine Thoracentesis Trainer
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Ready to master canine thoracocentesis?
  • Feel ribs, locate landmarks, and identify the optimal insertion site with a precise replica of the canine thorax
  • Practice draining air and fluid, replicating real-world conditions for accurate diagnosis and treatment
  • Durable and reusable cost-effective trainer is ideal for veterinary schools, clinics, and individual practitioners
  • Lightweight and intuitive, this portable and user-friendly trainer sets up and cleans up effortlessly for seamless training sessions
The Canine Thoracocentesis Trainer is your key to mastering this vital skill. This lifelike model replicates canine anatomy and pleural effusions, allowing you to hone your technique in a safe, controlled environment. Build precision, boost your confidence, and be prepared for real-world scenarios. Sharpen thoracocentesis skills without risk to animals or the need for cadavers. Gain confidence in performing this critical procedure successfully. Enjoy a cost-effective training tool that provides lasting value.
Caution: This product is a non-medical device intended solely for educational purposes. It is not designed, manufactured, or intended for use in the diagnosis, treatment, cure, mitigation, or prevention of disease or for any other medical purpose.