AquaSprouts® 29 Gallon Garden
Supplier: AQUASPROUTS, INC
84018
470357-160EA 349.95 USD
470357-160
AquaSprouts® 29 Gallon Garden
Educational Materials Environmental Educational Materials Environmental Learning Activities
Dive into the cutting-edge of sustainable education with the 29 gallon AquaSprouts® garden.
  • Durable birch frame and custom grow bed: Crafted from sustainably sourced birch wood, the frame fits securely over any standard 29-gallon aquarium; the sleek design integrates easily into any educational setting
  • 160 GPH submersible pump and Wi-Fi smart plug: System includes a powerful pump to maintain water circulation, with a smart plug that allows for easy control of water cycles via mobile app
  • Adjustable light bar: Removable lighting mounts allow a grow light to hang above the garden, and can be further extended to accommodate taller plants; light bar allows garden use anywhere, even without sufficient natural light
  • Porous clay grow media: Porous pebbles act as a mechanical filter and biofilter substrate, trapping fish waste and uneaten food while providing ample surface area for beneficial bacteria to grow and convert waste into nutrients (grow media does not degrade)
  • Interdisciplinary STEM learning: AquaSprouts® garden serves as a live, interactive platform for teaching a wide range of STEM concepts, from the basics of ecology and biology to the more complex principles of chemistry and physics
  • Hands-on experiments: Students can design and conduct experiments related to water quality, plant growth rates, and fish health, fostering critical thinking and scientific inquiry
  • Sustainability in action: System provides a concrete example of sustainable practices, emphasizing the importance of resource conservation and environmental responsibility
This comprehensive system is ideal for fostering hands-on learning in STEM subjects, allowing students to explore the interconnectedness of biological and environmental sciences in a captivating and engaging way.
The AquaSprouts® garden is more than just a garden - it's a fully functioning aquaponics ecosystem that repurposes a standard 29-gallon aquarium into an innovative, self-sustaining growing system. Here’s how it works: fish waste provides the nutrients that plants need to grow, while the plants in turn, purify the water. This closed-loop system not only illustrates the principles of ecology but also offers a real-world application of sustainability practices, making it an invaluable resource for educators aiming to inspire the next generation of environmental stewards.
Designed with simplicity and versatility in mind, the AquaSprouts® garden is suitable for classrooms, homes, or any educational setting where experiential learning is encouraged. Whether you’re teaching the nitrogen cycle, water chemistry, or plant biology, this system serves as a live laboratory, bringing textbook concepts to life and allowing students to observe, experiment, and learn through direct interaction.
Ordering information: 29-gallon aquarium and grow light are not included with the AquaSprouts® garden. These items can be purchased separately to complete an aquaponic setup.