Klinger Hall Effect Experiment
Supplier: Klinger
KSCIHLEF
470356-828EA 1650 USD
470356-828
Klinger Hall Effect Experiment
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A complete setup: everything you need to investigate the Hall effect.
  • Gauss meter and digital multimeter included
  • Power supplies included
The resistivity measurements of semiconductors cannot reveal whether one or two types of carriers are present; nor distinguish between them. However, this information can be obtained from Hall Coefficient measurements, which are also basic tools for the determination of carrier density and mobility in conjunctions with resistivity measurement.
When a conductor through which current is flowing, is placed in the magnetic field, a potential difference is generated between two opposite edges of the conductor in the direction mutually perpendicular to both the field and the conductor. This potential developed is Hall voltage and the phenomenon is called Hall effect. In the experimental setup, the crystal mounted on PCB is placed perpendicular to the pole pieces. A constant current is passed through the crystal using the constant current source. Magnetic field is produced by electromagnet operated by 0 to 12 V DC, 5 A power supply. Magnetic field intensity is measured by gauss meter with gauss probe. A Hall voltage thus produced is measured by the multimeter.
Ordering information: Kit includes (1) Hall probe, (1) Advanced power supply, (1) Electromagnet setup, (1) Constant current power supply, (1) Digital Gauss meter, (1) 3 ½ Digit multimeter.