Ward's® Mini Centrifuge
Supplier: Heathrow Scientific
121007
470356-704CS 1122 USD
470356-704
Ward's® Mini Centrifuge
Centrifuges Microcentrifuges
The Ward's® mini centrifuge is easy to use and brings increased efficiency to the laboratory. Tool-free snapspin rotors and quick-spin options make the centrifuge fast and easy to use.
  • Easy to use
  • 5 year warranty
The locking lid has been functionally designed to fit in the palm of the hand for easy repeat opening and quick tube extraction with one hand. Large lid size accommodates longer and skirted tubes and tall tube lids. Braking function quickly stops spinning when powered off or when the lid is opened. A weighted rubberized base eliminates potential movement and dampens vibration and noise. Ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (1.5/2.0, 0.5, 0.2 ml) for customized use.
Certifications: UL (Canada and US), ROHS3, CE, WEEE, UKCA and C-Tick.
Delivery information: Package includes the mini centrifuge, 6-place 1.5/2.0 ml tube rotor, 16-place 0.2 ml tube rotor (single or 2×8 strip), 6 tube adapters 0.2 ml, 6 tube adapters 0.5 ml, 4×O-rings for secure lid sealing, storage case for rotors and adapters, tube rack, low voltage (100 to 240 VAC, 50/60 Hz) double insulated power adapter with four interchangeable plugs.