Supertek® Resonance Apparatus
Supplier: Shiv Dial Sud & Sons
PH36178P
470356-598CS 459 USD
470356-598
Supertek® Resonance Apparatus
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
An attractive an easy to use resonance apparatus.
  • Tuning forks available separately
Investigate the length, frequency and velocity of sound waves by studying the resonance of air columns. A vibrating tuning fork, held over the top of the graduated water cylinder, will begin to resonate with the air column above the water in the cylinder. After taking the measurements of the air column and the tuning fork, the speed of sound can be calculated.