Lead(II) acetate trihydrate, single crystal substrate ACS
Supplier: ALDON CORP SE
|
Warning
|
Synonyms:
Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate
AD-LL0090-10KGLG
470356-494EA
668.4
USD
470356-494
Lead(II) acetate trihydrate, single crystal substrate ACS
Lead (II) acetate trihydrate
Formula:
(H₃CCOO)₂Pb·3H₂O MW: 379.34 g/mol |
MDL Number:
MFCD00150023 CAS Number: 6080-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...