Celestron FirstScope Signature Telescope
Supplier: Celestron International
22016
470356-312EA 100.95 USD
470356-312
Celestron FirstScope Signature Telescope
Telescopes
Celestron has joined forces with master lunar imager Robert Reeves on this addition to the beloved FirstScope family. The FirstScope Signature Series: Moon by Robert Reeves is a quality Dobsonian-style telescope that features a spherical glass mirror with a generous 76 mm of aperture.
  • The Signature Series FirstScope features a superb Moon image by astroimager and Team Celestron member Robert Reeves
  • High-quality tabletop Dobsonian with a 76 mm reflector optical tube
  • FirstScope artwork highlights key features on the lunar surface like craters and maria that can be explored with this telescope
  • Simple, portable design that’s easy to use for astronomers of all levels; a great choice for kids and families
  • Also includes a free PDF download of the Lunar Landscapes ebook by Robert Reeves
The FirstScope Signature Series: Moon by Robert Reeves telescope features a wide field-of-view which provides bright, sharp views of the Moon’s mountains and craters in crisp detail. Enjoy using the FirstScope to pan the Milky Way, explore open star clusters, marvel at Saturn’s rings, or view favorite deep-sky objects like the Orion Nebula.
This compact, lightweight telescope is an ideal entry-level astronomical telescope. Just remove it from the box, insert an eyepiece, and it is ready to view. To navigate the sky, simply rotate the azimuth axis in the direction of a desired object. It doesn’t get much simpler or more portable than the FirstScope. Its compact design makes the FirstScope Signature Series easy to take along on camping trips, picnics, or hikes. When not in use, the FirstScope Signature Series: Moon by Robert Reeves will be a stylish decorative fixture on a bookshelf or desk.
Explore the Moon:
Familiarize yourself with the Moon, our neighboring world. Robert Reeves’ signature FirstScope comes with a custom wrap that highlights 10 lunar features to explore including Tycho, Plato, and Copernicus. Learn more about the Moon with Robert Reeves’ Lunar Landscape eBook which goes into detail about the sizes of craters, how marias are formed, the natural history of the Moon, and other fascinating tidbits.
About Robert Reeves:
Robert Reeves first photographed the Moon in 1959, just as the space program was setting its sights on lunar exploration. Since then, he’s become one of the world’s best lunar imagers and a prolific author on astrophotography. Using Celestron telescopes from his home in Texas, Robert captures more detailed images than professional astronomers could during the heyday of the Apollo missions. To learn more information about Robert Reeves, check out his Team Celestron page or for Robert’s personal perspective on having his own telescope in the Signature Series.