OHAUS® Endeavor™ Microplate Shaker
Supplier: OHAUS CORP.
30770911
470356-212EA 1030 USD
470356-212
OHAUS® Endeavor™ Microplate Shaker
Shakers and Mixers Microplate Shakers
Stable, long-term shaking with triple eccentric drive and maintenance-free brushless DC motor.
  • Adaptable to a wide variety of accessories to customize for low to mid volume applications
  • Programmable with a simple on-board menu to save time and guarantee reproducible results
  • Remote function permits PC control and data transfer
For use in a wide variety of applications including Microbiology, Molecular Biology, Cell Cultures, Staining/Destaining, DNA Studies, Environmental and Food Sciences, Chemistry, General Mixing and more.
Feature a bright, backlit LCD display for speed and time. The timer will count up elapsed time or when set, timer will count down and shut off shaking when time reaches zero. A green indicator light illuminates when the unit is shaking with visual and audio alarm when timer counts down to zero or an error occurs.
Small footprint to save bench space, durable chemical-resistant polymer housing, simple knob control for timer and shaking, easy to navigate menu for advanced features.
Certifications: Product Safety: CE, TUV, TÜV SÜD certified to CAN/CSA C22.2 61010-1 ; CAN/CSA C22.2 61010-2-051; UL 61010-1; UL 61010-2-051.
Electromagnetic Compatibility: Canada ICES-003 Class B; FCC Part 15 Class B.