Engino® STEM Mechanics: Gears and Worm Drives
Supplier: United Scientific Supplies
STEM05
470356-050KT 40.03 USD
470356-050
Engino® STEM Mechanics: Gears and Worm Drives
Educational Materials Physics Educational Materials Mechanics Learning Activities
Design structures that use gears to change the force on an object to do work.
  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
Learn how gears can easily reduce or increase speed, change force or transfer motion from one position to another. Discover how worm drives are used to greatly reduce rotational speed and how screws can convert rotational motion to linear, while greatly increasing force. Build 12 working models such as an experimental crane, a gearbox, a carousel, a helicopter, a screw press and a crane with a rotating arm.
You can find easy-to-follow building instructions for all models either online or in the booklet included. The booklet provides detailed explanations of the different scientific principles applied and incorporates innovative experimental activities for hands-on learning. A Quiz section is also available to challenge your newly acquired knowledge!