Engino® STEM Simple Machines Set
Supplier: United Scientific Supplies
UNE961
470356-038KT 62.21 USD
470356-038
Engino® STEM Simple Machines Set
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Explore the engineering advantage of different simple machines.
  • Download easy-to-follow instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
This set covers the subject of simple machines in an encompassing and fascinating way. These mechanisms offer a mechanical advantage and are crucial elements of many machines and devices. They are designed to multiply or reduce force, increase or decrease speed and convert one type of motion to another.
The set includes enough parts to create 60 working models that cover the subjects of levers, the wedge, the wheel and axle, the screw, the inclined plane, the pulley, as well as the more advanced gears and linkages! You can find easy-to-follow building digital instructions for all models along with detailed explanations of the different scientific principles applied. The methodology suggested combines theory with innovative experimental activities that lead to hands-on learning and engineering creativity.
Suggested for students working in groups of 2 or 3.
Delivery information: Kit includes 191 plastic parts, 925 connecting points, digital instructions for 60 models, gearing system, pulleys system.