United Scientific Laboratory Scoops, PP, 500 ml
Supplier: United Scientific Supplies
81258PK160
470355-710CS 543.58 USD
470355-710
United Scientific Laboratory Scoops, PP, 500 ml
Scoops
Ergonomically designed scoops for transferring powders.
  • Polypropylene construction
  • Will not rust or corrode
  • Autoclavable
These sturdy polypropylene scoops make sampling powders easier to manage and transfer. The capacity is molded into the top of each scoop for easy reference. The polypropylene material is autoclavable and will not rust or corrode.