Digital Anemometer
Supplier: American Scientific
6577-01
470355-688EA 30.5 USD
470355-688
Digital Anemometer
Anemometers
Elevate your weather monitoring with our 4-in-1 handheld digital anemometer.
  • Clear, easy-to-read screen showing wind speed and other relevant data
  • Measures wind speed in various units such as mph, km/h, m/s, and knots
  • Provides precise wind speed measurements for accurate readings
  • Compact and lightweight design, making it easy to carry and use in the field
  • Essential for meteorologists and weather enthusiasts to monitor and record wind conditions accurately
  • Useful for outdoor activities such as sailing, flying drones, and paragliding, where knowing wind speed is critical for safety
  • Intuitive interface and easy-to-read display make it accessible for users of all skill levels
The American Scientific digital anemometer delivers precise wind speed. Versatile and compact, it suits classroom research, drone pilots, HVAC techs, sailors, and more. The backlit LCD screen displays readings in multiple units, with a MAX mode and data memory function. Durable and portable, it comes with a protective case. This digital anemometer will truly enhance your weather/wind measurements!