Pendulum Wave Demo
Supplier: American Scientific
3645-00
470355-428EA 99 USD
470355-428
Pendulum Wave Demo
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Discuss the dependence of a pendulum's period on its length with this beautiful wave demo.
  • Teacher's guide included
  • Easily assembled in minutes
This item makes a captivating demonstration for all ages! The Pendulum Wave Demo simulates phases, beats, and frequencies. It features a stand with nine (9) 22 mm (0.88") steel ball pendulums and comes with an initiator stick, which helps to simultaneously release all pendulums. When released, the pendulums will swing back and forth to quickly form a wave pattern and then cycle through different phases. This product is great for a classroom, as a gift, or for display.