Ferrofluid Magnetic Display
Supplier: American Scientific
3486-00
470355-412EA 35.7 USD
470355-412
Ferrofluid Magnetic Display
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Visualize magnetic fields or discuss advanced materials.
  • Not a toy: intended for educational use only, ages 12 and up
  • Teacher's guide included
Ferrofluid is the astonishing liquid that becomes highly magnetized in the presence of a magnetic field. Comprised of nano-scale magnetic particles suspended in a transparent liquid, undisturbed Ferrofluid forms into a smooth pool. But when a magnet is brought close, Ferrofluid reacts in strange and fascinating ways. With Ferrofluid in a bottle, you can safely and easily play with this spectacular substance. Using two included neodymium magnets, you can spike, toss, drop, pull, and bridge the Ferrofluid. You can even make it dance.