RNA Viral Infection RT-PCR Kit
Supplier: GENOSENSOR CORPORATION
3401
470355-108EA 223 USD
470355-108
RNA Viral Infection RT-PCR Kit
Educational Materials Biology Educational Materials PCR Learning Activities
The RNA virial infection RT-PCR kit is designed to introduce students to the RNA viral world and RT-PCR – an RNA amplification technology.
  • Perform an RNA amplification via RT-PCR technique
  • Verify presence of RT-PCR amplified RNA by gel electrophoresis
  • Fast and simple protocol for easy integration into biotechnology lessons
In this lab, students play the role of the diagnostician in the following scenario. Patients arrive at the emergency room with fever and cough. An RNA viral infection is suspected and must be confirmed or ruled-out from patient samples. A short RNA is used to emulate the virus; no actual viral particles are used.