miniPCR® DNA Discovery System™ Pro
Supplier: AMPLYUS LLC
QP-2020-16
470355-084EA 2299.99 USD
470355-084
miniPCR® DNA Discovery System™ Pro
Educational Classroom Kits and Activities
Your professional biotech toolkit.
  • 16×0.2 ml PCR tube capacity (strip-compatible)
  • Bluetooth® control from your phone or laptop
  • Fully compatible with standard tubes and reagents
  • Integrated blue-light transilluminator
  • Compatible with SeeGreen™, GelGreen®, SYBR® and other blue-excitable dyes
  • Adjustable voltage and timer function
  • Compatible with gel excision and downstream applications
The miniPCR DNA Discovery System™ Pro includes a miniPCR® mini16X thermal and a GELATO™ electrophoresis and visualization system.
Ordering information: Backed by a 2-year warranty.