Wallcur® Practi-Ipratropium Bromide Nebules
Supplier: WALLCUR, LLC.
1206PN
470355-072BX 53.75 USD
470355-072
Wallcur® Practi-Ipratropium Bromide Nebules
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Made for training.
  • Contains no medication
  • Contents are safe for use with most simulators
Practi-Ipratropium Bromide™ 0.02% nebules simulates 0.5 mg/2.5 ml of ipratropium bromide 0.02% (Atrovent®) in a nebulizer. Practi-ipratropium comes as a liquid solution that is to be inhaled by the patient's mouth using a special nebulizer. Safely teach this skill using Wallcur's Practi-ipratropium bromide 0.02% with any task trainer or manikin that accepts distilled water. Students can also practice dosage calculations to understand the different dosages for adults versus pediatrics.
Delivery information: These products require a release form for purchase. If your account does not have one on file, the item cannot ship until release documentation is received.
Caution: For training purposes only. Not for human or animal use.