Wallcur® Practi-Insulin Pen Trainer
Supplier: WALLCUR, LLC.
1520PN
470354-934EA 23.2 USD
470354-934
Wallcur® Practi-Insulin Pen Trainer
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Practice insulin injection using an insulin pen.
  • Contains no medication or needle
Wallcur's Practi-Insulin Pen Trainer™ for clinical training, is a 3 ml (100 units/ml) simulated disposable insulin pen with a dose selection dial and a total liquid fill of 300 units. Provide students with a safe, simulated practice device in the correct method of administering medication from a pre-loaded insulin auto-injector system. An essential tool for teaching diabetes management, the Practi-Insulin Pen Trainer teaches the advantages of insulin self-administration and is adjustable from 0 to 60 units with a calibration window for precise dose verification.
Delivery information: These products require a release form for purchase. If your account does not have one on file, the item cannot ship until release documentation is received.
Caution: For training purposes only. Not for human or animal use.