Shaking Water Bath
Supplier: Ward's
SB0012
470354-886EA 3906 USD
470354-886
Shaking Water Bath
Baths Shaking Water Baths
Included spring platform accepts tubes, bottles and flasks.
  • Orbital shaking motion, 30 to 200 rpm
  • Temperature range: Ambient +5 to 80 °C
  • Digital display (time/temperature/speed)
This compact 12 L shaking water bath has precise temperature control up to 80 °C in 0.1 °C increments. The built-in digital timer can be set from 1 minute to 48 hours, or continuous operation. Shaking orbit is 25 mm, temperature accuracy/uniformity is 0.3/0.2 °C. The included spring platform can accommodate 15/50 ml centrifuge tubes, Erlenmeyer flasks and media bottles. The included hinged lid is also removable for complete versatility.